Learn More
Bioworld HUMAN MONOCYTE CHEMOTACTIC
Catalog No. 50253662
Encompass_Preferred
HUMAN MONOCYTE CHEMOTACTIC PROTEIN-3 (CCL7), (RECOMBINANT): Recombinant Human Monocyte Chemotactic Protein-3 (CCL7)- MCP-3 Source:E.Coli Description : Recombinant Human MCP-3 produced in E.Coli is a non-glycosylated, Polypeptide chai...
Catalog No. 50-253-662 Supplier Bioworld Supplier No. 220605622
HUMAN MONOCYTE CHEMOTACTIC PROTEIN-3 (CCL7), (RECOMBINANT): Recombinant Human Monocyte Chemotactic Protein-3 (CCL7)- MCP-3 Source:E.Coli Description : Recombinant Human MCP-3 produced in E.Coli is a non-glycosylated, Polypeptide chai...