Learn More
Biomatik Corporation Human Monocyte Chemotactic Protein 3 (MCP3) CLIA Kit
Supplier:  Biomatik Corporation EKU08820
Encompass_Preferred
Human Monocyte Chemotactic Protein 3 (MCP3) CLIA Kit for detecting MCP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.26pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P80098. Gene ID: 6354. Target Synonyms: CCL7; FIC; MARC; NC28; MCP3; SCYA7; Chemokine C-C-Motif Ligand 7; Small Inducible Cytokine A7; Monocyte Chemoattractant Protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8621