Learn More
Sino Biological Recombinant Human, Mouse, Rat, Rhesus, Canine BMP-2 Protein
Supplier:  Sino Biological 10426HNAE120
Encompass_Preferred
A DNA sequence encoding the mature form of human / mouse / rat / rhesus / canine BMP2 (NP_001191.1) (Ala284-Arg396) was expressed with an initial Met. The mature form sequences of human, mouse, rat, rhesus and canine BMP2 are identical.
Catalog No. 50-255-5307