Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Mouse FGF-8b Recombinant Protein, PeproTech®

Catalog No. 100255UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-25-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 100-25-5UG Supplier Gibco™ Supplier No. 100255UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). Recombinant Human/Murine FGF-8 (FGF-8b) is a 22.5 kDa protein consisting of 194 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37237, P55075
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 14179, 2253
Molecular Weight (g/mol) 22.5 kDa
Name Human/Mouse FGF-8b
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Fibroblast Growth Factor 8
Common Name FGF-8B
Gene Symbol FGF8
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
Content And Storage -20°C
Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.