Learn More
Sino Biological Human IL23A/p19 Neutralizing Antibody
Supplier:  Sino Biological CT035MH066200
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the Human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Human IL23A & Mouse IL12B Heterodimer. The antibody was produced using recombinant antibody technology.
Catalog No. 50-237-3174