Learn More
Gibco™ Human/Mouse/Rat Activin A Recombinant Protein, PeproTech®
Description
120-14P-1MG will be provided as 2 x 500 μg (120-14P-500UG). Recombinant Human/Murine/Rat Activin A is a 26.0 kDa disulfide-linked homodimer of two betaA chains, each containing 116 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P08476, P18331, Q04998 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 16323, 29200, 3624 |
| Molecular Weight (g/mol) | 26 kDa |
| Name | Human/Mouse/Rat Activin A |
| Purification Method | Purified |
| Quantity | 10 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.