Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Mouse/Rat Activin A Recombinant Protein, PeproTech®

Catalog No. 1201410UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-14-10UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 120-14-10UG Supplier Gibco™ Supplier No. 1201410UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

120-14-1MG will be provided as 2 x 500 μg (120-14-500UG). Recombinant Human/Murine/Rat Activin A is a 26.0 kDa disulfide-linked homodimer of two betaA chains, each containing 116 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476, P18331, Q04998
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 16323, 29200, 3624
Molecular Weight (g/mol) 26 kDa
Name Human/Mouse/Rat Activin A
Purification Method Purified
Quantity 10 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Common Name Activin A
Gene Symbol Inhba
Biological Activity Determined by its ability to inhibit the proliferation of mouse MPC-11 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Content And Storage -20°C
Activity Determined by its ability to inhibit the proliferation of mouse MPC-11 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.