Learn More
Sino Biological Anti-BDNF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50240RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human / Mouse / Rat / Cynomolgus / Canine BDNF (rM BDNF; Catalog#50240-MNAS;P21237-1; Met1-Arg249). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3348