Learn More
Gibco™ Human/Mouse/Rat BMP-2 Recombinant Protein, PeproTech®
Description
Recombinant Human/Murine/Rat BMP-2 is a 26.0 kDa homodimeric protein consisting of two 115 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P12643, P21274, P49001 |
| For Use With (Application) | Bioactivity, Control, Western Blot Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 12156, 29373, 650 |
| Molecular Weight (g/mol) | 26 kDa |
| Name | Human/Mouse/Rat BMP-2 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.