Learn More
Sino Biological Anti-BMP-2 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10426MM03H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human / Mouse / Rat / Rhesus / Canine BMP-2 (rh BMP-2 ; Catalog#10426-HNAE; NP_001191.1; Gln283-Arg396). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8707