Learn More
Gibco™ Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®
Description
120-11-1MG will be provided as 2 x 500 μg (120-11-500UG). Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | O95390, Q9Z1W4, Q9Z217 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 10220, 14561, 29454 |
| Molecular Weight (g/mol) | 25 kDa |
| Name | Human/Mouse/Rat GDF-11 |
| Purification Method | Purified |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.