Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®

Catalog No. 1201150UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-11-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 120-11-50UG Supplier Gibco™ Supplier No. 1201150UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

120-11-1MG will be provided as 2 x 500 μg (120-11-500UG). Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95390, Q9Z1W4, Q9Z217
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 10220, 14561, 29454
Molecular Weight (g/mol) 25 kDa
Name Human/Mouse/Rat GDF-11
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11
Common Name GDF11
Gene Symbol GDF11
Biological Activity Assay 1: Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 0.08-0.10 ug/ml Assay 2: Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1cells. The ED50 for this effect is 8-10 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
Content And Storage -20°C
Activity Assay 1: Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 0.08-0.10 ug/ml. Assay 2: Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1cells. The ED50 for this effect is 8-10 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.