Learn More
Sino Biological Recombinant Human, Mouse, Rat, Rhesus, Canine BMP-2 Protein (hFc Tag) 100 µg
Supplier:  Sino Biological 10426H01H100
Encompass_Preferred
A DNA sequence encoding the mature form of human / mouse / rat / rhesus / canine BMP2 (NP_001191.1) (Gln 283-Arg 396) was fused with the Fc region of human IgG1 at the N-terminus. The mature form sequences of human, mouse, rat, rhesus and canine BMP2 are identical.
Catalog No. 50-224-7020