Learn More
STEMCELL Technologies Human/Mouse Recombinant NT-3, Size: 1000 ug
SDP
Supplier:  STEMCELL Technologies 78074.2
Encompass
Use neurotrophin-3 (NT-3) to generate -tubulin III peripheral neurons from neural crest stem cells (Menendez et al. Nat Protoc 2013) and oligodendrocyte precursor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells (Douvaras et al. Stem cell reports 2014). NT-3 is a member of the nerve growth factor (NGF) family of proteins that includes neuron growth factor (NGF) brain-derived neurotrophic factor (BDNF) and neurotrophin-4/5. NT-3 signals a number of trophic effects through its transducing receptor tyrosine kinase TrkC and is known to promote the survival development and differentiation of neurons (Chalazonitis. Mol Neurobiol 1996). Human/mouse recombinant NT-3 from STEMCELL comes lyophilized with ≥ 95% purity specific activity EC50 20 ng/mL (for Lot 1000050644 or higher) and EC50 20 μg/mL (for other lots) endotoxin levels verified 1 EU/μg protein. Alternative names HGNF
Catalog No. 50-239-0128
Add to Cart
Request a Quote