Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MP68 Isoform 2 (aa 1-24) Control Fragment Recombinant Protein

Catalog No. RP100879
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100879 Supplier Invitrogen™ Supplier No. RP100879
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63868. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MP68 Isoform 2 is a variant of the MP68 gene, which is known for its involvement in cellular processes related to structural maintenance and function in various tissue types. The MP68 gene, like many other genes, undergoes alternative splicing, leading to the production of different isoforms that can have distinct biological roles. Isoform 2 specifically is expressed predominantly in neuronal tissues, where it plays a critical role in the regulation of axonal growth and neuronal connectivity. The protein encoded by MP68 Isoform 2 is part of a larger family of structural proteins that contribute to the cytoskeleton's integrity, ensuring proper cell shape and intracellular transport. In addition to its structural role, MP68 Isoform 2 is involved in intracellular signaling pathways that influence cell survival and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Name Human MP68 Isoform 2 (aa 1-24) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias MP68
Common Name MP68 Isoform 2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MKLYGTRGSDISHFQCQMLQSIIK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.