Learn More
Sino Biological Human MPO PAb 100ul (Sino Biological)
Supplier:  Sino Biological 110728T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Myeloperoxidase/MPO, and purified by antigen affinity chromatography.
Catalog No. 50-237-3283