Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human mPR alpha (aa 1-72) Control Fragment Recombinant Protein

Catalog No. RP99833
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99833 Supplier Invitrogen™ Supplier No. RP99833
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61376. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The steroid progesterone induces the resumption of maturation in oocytes via a nongenomic pathway through binding to a novel, membrane progestin receptor (mPR). This pathway inhibits adenylyl cyclase and reduces intracellular cAMP, and also activates mitogen-activated protein kinase to effect signal transduction pathways. Three distinct groups, designated α, β and γ, comprise this gene family. mPRα, also designated progestin and adipoQ receptor family member VII (PAQR7), consists of an extracellular N-terminus, an intracellular C-terminus, and seven transmembrane domains. It is expressed in ovary, testis, placenta, uterus and bladder. mPRβ, also designated progestin and adipoQ receptor family member VIII (PAQR8), consists of eight putative transmembrane regions and an intracellular N-terminus that contains a leucine-rich motif. It is a 354 amino acid protein with a molecular mass of about 40 kDa and is expressed in brain and spinal cord. Both mPRα and mPRβ may be G protein-coupled receptors and may be involved in oocyte maturation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86WK9
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 164091
Name Human mPR alpha (aa 1-72) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2310021M12Rik; AV001994; BB007237; Membrane progesterone P4 receptor alpha; Membrane progesterone receptor alpha; Membrane progestin receptor alpha; mPR; mPR alpha; Mpra; MRPA; mSR; PAQR7; Pglp; PPAR-gamma-induced liver protein; Progesterone and adipoQ receptor family member 7; Progestin and adipoQ receptor family member 7; progestin and adipoQ receptor family member VII
Common Name mPR alpha
Gene Symbol PAQR7
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAMAQKLSHLLPSLRQVIQEPQLSLQPEPVFTVDRAEVPPLFWKPYIYAGYRPLHQTWRFYFRTLFQQHNEA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.