Learn More
BIOHIPPO Human MPZ Microsample
SDP
Catalog No. 5000315411
Encompass
Human MPZ Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MPZ in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MPZ Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MPZ Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MPZ biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15411 Supplier BIOHIPPO Supplier No. BHE15211453
Human MPZ Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MPZ in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MPZ Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MPZ Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MPZ biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color