Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MR-ProADM ELISA Kit

Catalog No. EEL189
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL189 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL189 Supplier Invitrogen™ Supplier No. EEL189
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Mid regional pro adrenomedullin ELISA Kit (MR ProADM) ELISA quantitates MR ProADM in serum, plasma and other biological fluids. Principle of the method The Human Mid regional pro adrenomedullin (MR ProADM) solid phase sandwich ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target specific antibody has been pre coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

MR-proADM, short for mid-regional proadrenomedullin, is a precursor protein that gives rise to the adrenomedullin (ADM) hormone. It is involved in various physiological processes, particularly in the regulation of cardiovascular function and inflammation. Functionally, MR-proADM is associated with vasodilation, blood pressure regulation, and immune response modulation. It acts as a biomarker for endothelial dysfunction and can be used to assess the severity of certain cardiovascular diseases. Elevated levels of MR-proADM have also been linked to systemic inflammation and sepsis. Structurally, MR-proADM is a protein that consists of a chain of amino acids. It undergoes post-translational processing to generate the active form of ADM, which is a shorter peptide consisting of 52 amino acids. Localization-wise, MR-proADM is primarily produced in various tissues, including the adrenal glands, lungs, and endothelial cells lining blood vessels. It is released into the bloodstream and can be measured as a biomarker to provide insight into cardiovascular health and inflammatory conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35318
Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Mid-regional proadrenomedullinMid-regional proadrenomedullin;
Gene ID (Entrez) 133
Gene Symbol ADM
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 100 μL, Plasma 100 μL, Serum 100 μL, Tissue Homogenate 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target MR-proADM
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.