Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human MRGPRX2 Full-Length Protein (VLP), HEK293-expressed, 100 μg

Catalog No. NC3874396
Encompass_Preferred

Full-length human MRGPRX2 presented as virus-like particles (VLPs), expressed in HEK293 cells. The VLP format displays the receptor in a membrane-like context suitable for antibody development, immunization, and receptor interaction studies, with low endotoxin and defined particle size.

  • Expression system: Human 293 (HEK293) cells.
  • Amino acid range: Asp 2-Val 330 (Accession Q96LB1).
  • Product format: Virus-like particle (VLP).
  • Available sizes: 20 μg, 100 μg, 500 μg (250 μg x 2).
  • Endotoxin level: <1.0 EU/μg (LAL/rFC).
  • Particle size: Mean peak radius 60-80 nm by DLS.
  • Storage buffer: PBS with arginine, pH 7.4, with trehalose as protectant.
  • Storage conditions: Store at -70°C; avoid repeated freeze-thaw cycles.

Catalog No. NC3874396 Supplier ACROBiosystems Supplier No. MR2H52P3100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Full-length human MRGPRX2 presented as virus-like particles (VLPs), expressed in HEK293 cells. The VLP format displays the receptor in a membrane-like context suitable for antibody development, immunization, and receptor interaction studies, with low endotoxin and defined particle size.

  • Expression system: Human 293 (HEK293) cells.
  • Amino acid range: Asp 2-Val 330 (Accession Q96LB1).
  • Product format: Virus-like particle (VLP).
  • Available sizes: 20 μg, 100 μg, 500 μg (250 μg x 2).
  • Endotoxin level: <1.0 EU/μg (LAL/rFC).
  • Particle size: Mean peak radius 60-80 nm by DLS.
  • Storage buffer: PBS with arginine, pH 7.4, with trehalose as protectant.
  • Storage conditions: Store at -70°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.