Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MRP7 (aa 769-865) Control Fragment Recombinant Protein

Catalog No. RP94747
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94747 Supplier Invitrogen™ Supplier No. RP94747
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61008. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The MRP family is represented by nine similar ABC transporters that have the ability to transport structurally diverse lipophilic anions and operate as chemical efflux pumps. MRP7 (multidrug resistance-associated protein 7, ATPbinding cassette sub-family C member 10) is a multi-pass membrane protein that belongs to the ABC transporter family (conjugate transporter subfamily). MRP7 is involved with the ATP-dependent transport of 17β-estradiol-D-17-β-glucuronide (E217βG). MRP7 is also probably involved in cellular detoxification through its lipophilic anion extrusion capabilities. MRP7 contains two ABC transmembrane type 1 domains and two ABC transporter domains. MRP7 likely has three isoforms. Isoform 2 is the most widely expressed, while isoform 1 is predominately expressed in the spleen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5T3U5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 89845
Name Human MRP7 (aa 769-865) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ABCC10; ATP binding cassette subfamily C member 10; ATP-binding cassette sub-family C member 10; ATP-binding cassette, subfamily C (CFTR/MRP), member 10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; EST182763; mFLJ00002; Mrp7; Multidrug resistance-associated protein 7; SIMRP7
Common Name MRP7
Gene Symbol ABCC10
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ILGMLSYTTRLLCTHRTEYLERADAVLLMEAGRLIRAGPPSEILPLVQAVPKAWAENGQESDSATAQSVQNPEKTKEGLEEEQSTSGRLLQEESKKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.