Learn More
Sino Biological Anti-MRPL13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 203792T44100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human MRPL13 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1699