Learn More
Sino Biological Human MS4A1 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 11007R001P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, Human CD20 Transfected CHO Cells and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-3634