Learn More
As One International Inc Human Mesenchymal Stem Cell Growth Medium, 1kit
Catalog No. 501654996
Encompass_Preferred
Human Mesenchymal Stem Cell Growth Medium is developed for the optimal proliferation of human mesenchymal stem cells, allowing them to expand through multiple passages while retaining the ability to differentiate into osteoblasts, adipocytes, and chondrocytes.
Catalog No. 50-165-4996 Supplier As One International Inc Supplier No. HUXMX90011
Human Mesenchymal Stem Cell Growth Medium is developed for the optimal proliferation of human mesenchymal stem cells, allowing them to expand through multiple passages while retaining the ability to differentiate into osteoblasts, adipocytes, and chondrocytes.