Learn More
Sino Biological Recombinant Human Mesothelin Protein (AF 647 site-specific conjugation, His Tag)
Catalog No. 502993206
Encompass_Preferred
A DNA sequence encoding the human MSLN (AAH09272.1) (Glu296-Gly580) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 647 (Excitation Max.= 655 nm, Emission Max.= 680 nm).
Catalog No. 50-299-3206 Supplier Sino Biological Supplier No. 13128H86H1SG100
A DNA sequence encoding the human MSLN (AAH09272.1) (Glu296-Gly580) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 647 (Excitation Max.= 655 nm, Emission Max.= 680 nm).