Learn More
BIOHIPPO Human MSLN Microsample
SDP
Catalog No. 5000314562
Encompass
Human MSLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSLN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MSLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MSLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MSLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14562 Supplier BIOHIPPO Supplier No. BHE15211825
Human MSLN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSLN in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MSLN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MSLN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MSLN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color