Learn More
Sino Biological Recombinant Human Mesothelin Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 13128H08HB100
Encompass_Preferred
A DNA sequence encoding the Human MSLN (Q13421-2) (Met1-Arg286) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4312