Learn More
Sino Biological Recombinant Human Mesothelin Protein, HPLC-verified
Supplier:  Sino Biological 13128HNCH500
Encompass_Preferred
A DNA sequence encoding the human MSLN (NP_005814.2) (Glu296-Gly580) was expressed with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-255-4311