Learn More
Sino Biological Anti-IGBF/PRSP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106783T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IGBF/PRSP, and purified by antigen affinity chromatography.
Catalog No. 50-240-8339