Learn More
BIOHIPPO Human MSN Microsample
SDP
Catalog No. 5000314938
Encompass
Human MSN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSN in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MSN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MSN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MSN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14938 Supplier BIOHIPPO Supplier No. BHE15211617
Human MSN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSN in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MSN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MSN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MSN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color