Learn More
ACROBiosystems Human MSP1D1 30ug
Catalog No. 502535494
Encompass_Preferred
Biotinylated Human MSP1D1 Protein HisAvitag is expressed from E. coli cells. It contains AA Ser 1- Gln 211.
Catalog No. 50-253-5494 Supplier ACROBiosystems Supplier No. APOH81Q530UG
Biotinylated Human MSP1D1 Protein HisAvitag is expressed from E. coli cells. It contains AA Ser 1- Gln 211.