Learn More
Sino Biological Human MSR1 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372932
Encompass_Preferred
Each vial contains 48 ng of recombinant Human MSR1 / CD204. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-2932 Supplier Sino Biological Supplier No. SEK104275
Each vial contains 48 ng of recombinant Human MSR1 / CD204. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 5000 pg/mL is recommended.