Learn More
Sino Biological Anti-RON/MSP receptor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11608MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD136 / MST1R (rh CD136 / MST1R; Catalog#11608-H08H; Q04912; Met 1-Leu 571). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7861