Learn More
Sino Biological Anti-RON/MSP receptor Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11608MM03P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD136/MST1R (rh CD136/MST1R; Catalog#11608-H08H; Q04912; Met1-Leu571) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2260