Learn More
BIOHIPPO Human MT Microsample
SDP
Catalog No. 5000313875
Encompass
Human MT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MT in tissue homogenates cell lysates and other biological fluids It provides a detection range of 4 69-300 pg/mL with a sensitivity of 2 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13875 Supplier BIOHIPPO Supplier No. BHE15210572
Human MT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MT in tissue homogenates cell lysates and other biological fluids It provides a detection range of 4 69-300 pg/mL with a sensitivity of 2 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color