Learn More
BIOHIPPO Human MTOR Microsample
SDP
Catalog No. 5000315350
Encompass
Human MTOR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MTOR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MTOR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MTOR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MTOR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15350 Supplier BIOHIPPO Supplier No. BHE15210559
Human MTOR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MTOR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MTOR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MTOR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MTOR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color