Learn More
Sino Biological Human MUC-1 Protein (aa890-1158, His Tag), Biotinylated
Catalog No. 502992787
Encompass_Preferred
A DNA sequence encoding the Human MUC1 (P15941-1) (Ser890-Gly1158) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-299-2787 Supplier Sino Biological Supplier No. 30117H08HB100
A DNA sequence encoding the Human MUC1 (P15941-1) (Ser890-Gly1158) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.