Learn More
Sino Biological Anti-MUC-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12123RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Mucin-1 / MUC-1 (rh MUC1; NP_001018016.1;Met1-Gly167). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8310