Learn More
Sino Biological Recombinant Human MUC-1 Protein (His Tag)
Supplier:  Sino Biological 30117H08H1100
Encompass_Preferred
A DNA sequence encoding the Human MUC1 (P15941-1) (Met1-Ser380) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-4259