Learn More
Sino Biological Recombinant Human MUC17 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 30134H08H100
Encompass_Preferred
A DNA sequence encoding the Human MUC17 (Q685J3-1) (Arg4131-Leu4390) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3801