Learn More
BIOHIPPO Human MUC5B Microsample
SDP
Catalog No. 5000314621
Encompass
Human MUC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC5B in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MUC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MUC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MUC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14621 Supplier BIOHIPPO Supplier No. BHE15212114
Human MUC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC5B in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human MUC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human MUC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human MUC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color