Learn More
ACROBiosystems Human Mucin-1 100ug
Catalog No. 502534409
Encompass_Preferred
Human Mucin-1 (24-380) Protein His Tag (MU1-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 380 (Accession # P15941-1).
Catalog No. 50-253-4409 Supplier ACROBiosystems Supplier No. MU1H52H7100UG
Human Mucin-1 (24-380) Protein His Tag (MU1-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 380 (Accession # P15941-1).