Learn More
ACROBiosystems Human Mucin-1 / MUC-1 (24-380) Protein, Fc Tag (MALS verified)
Catalog No. 502055295
Encompass_Preferred
Recombinant Protein;1MG;Human Mucin-1 (24-380), Fc Tag (MU1-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 380 (Accession # P15941-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human Mucin-1 (24-380), Fc Tag (Cat. No. MU1-H5254) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Mucin-1 Antibody, Mouse IgG1 (SM-3) with a linear range of 0.5-4 ng/mL (QC tested).;Immobilized Monoclonal Anti-Mucin-1 Antibody, Mouse IgG1 (SM-3) at 2 μg/mL (100 μL/well) can bind Human Mucin-1 (24-380), Fc Tag (Cat. No. MU1-H5254) with a linear range of 0.2-2 ng/mL (Routinely tested).
Catalog No. 50-205-5295 Supplier ACROBiosystems Supplier No. MU1H52541MG
Recombinant Protein;1MG;Human Mucin-1 (24-380), Fc Tag (MU1-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 380 (Accession # P15941-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human Mucin-1 (24-380), Fc Tag (Cat. No. MU1-H5254) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Mucin-1 Antibody, Mouse IgG1 (SM-3) with a linear range of 0.5-4 ng/mL (QC tested).;Immobilized Monoclonal Anti-Mucin-1 Antibody, Mouse IgG1 (SM-3) at 2 μg/mL (100 μL/well) can bind Human Mucin-1 (24-380), Fc Tag (Cat. No. MU1-H5254) with a linear range of 0.2-2 ng/mL (Routinely tested).