Learn More
ACROBiosystems ACROBiosystems Human Mucin-1 / MUC-1 (116-173) Protein, Fc Tag
Catalog No. 502107294
Encompass_Preferred
Recombinant Protein;1MG;Human Mucin-1 (116-173), Fc Tag (MU1-H5153) is expressed from E.coli cells. It contains AA Ser 116 - Gly 173 (Accession # P15941-8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.7 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-7294 Supplier ACROBiosystems Supplier No. MU1H51531MG
Recombinant Protein;1MG;Human Mucin-1 (116-173), Fc Tag (MU1-H5153) is expressed from E.coli cells. It contains AA Ser 116 - Gly 173 (Accession # P15941-8).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.7 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).