Learn More
ACROBiosystems Human Mucin-13 100ug
Catalog No. 502534406
Encompass_Preferred
Human Mucin-13 Protein His Tag (MU3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Leu 421 (Accession # Q9H3R2).
Catalog No. 50-253-4406 Supplier ACROBiosystems Supplier No. MU3H52H3100UG
Human Mucin-13 Protein His Tag (MU3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Leu 421 (Accession # Q9H3R2).