Learn More
Biomatik Corporation Human Mucin 17 (MUC17) ELISA Kit
Supplier:  Biomatik Corporation EKU06019
Encompass_Preferred
Human Mucin 17 (MUC17) ELISA Kit for detecting MUC17 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q685J3. Gene ID: 140453. Target Synonyms: MUC3; Mucin Cell Surface Associated; Small intestinal mucin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7684