Learn More
Biomatik Corporation Human Multidrug Resistance Associated Protein 1 (MRP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12164
Encompass_Preferred
Human Multidrug Resistance Associated Protein 1 (MRP1) ELISA Kit for detecting MRP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33527. Gene ID: 4363. Target Synonyms: ABCC1; ABC29; ABCC; GS-X; ATP-Binding Cassette,Subfamily C (CFTR/MRP) Member 1; Leukotriene C (4) transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8017