Learn More
Cellular Engineering Technologies Cryopreserved vial of human multipotent iPS cells containing ≥ 500,000 cells
Supplier:  Cellular Engineering Technologies CR1002500
Encompass_Preferred
Cells isolated from cord blood containing lower MHC Class II molecules are programmed using our patented virus-free method using episomal DNA and our proprietary mix of vectors, excluding l-Myc, c-Myc, and Lin28 transcription factors.The cell line was validated for pluripotency based on colony morphology, alkaline phosphatase expression, and expression of SSEA-4. Cells are free of Mycoplasma and exhibit classical morphology and growth characteristics. Multipotent stem cells can self-renew through dividing and can be developed into multiple specific cell types for a particular tissue tropism. These cells can be differentiated into all cell types within one lineage for cell therapy, regenerative medicine, and fertility applications.Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753004