Learn More
American Research Products Inc Human Muscle Skeletal Receptor
Catalog No. 5000328632
Encompass
Human Muscle Skeletal Receptor Tyrosine Kinase (MUSK) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28632 Supplier American Research Products Inc Supplier No. CSBE15804H
Human Muscle Skeletal Receptor Tyrosine Kinase (MUSK) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich