Learn More
Biomatik Corporation Human MutY Homolog (MUTYH) ELISA Kit
Supplier:  Biomatik Corporation EKU06046
Encompass_Preferred
Human MutY Homolog (MUTYH) ELISA Kit for detecting MUTYH in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UIF7. Gene ID: 4595. Target Synonyms: MYH; A/G-specific adenine DNA glycosylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5104